Assay Details
Target Gene Details
Entrez Gene ID: | 1261 |
Gene Name: | cyclic nucleotide gated channel alpha 3 |
Gene Aliases: |
ACHM2, CCNC1, CCNCa, CCNCalpha, CNCG3, CNG3 |
Location: |
Chr.2:98346155-98399154 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNGA3 | NM_001079878.1 | NP_001073347.1 | ||
| NM_001298.2 | NP_001289.1 | |||
| XM_006712243.2 | XP_006712306.1 | |||
| XM_011510554.2 | XP_011508856.1 | |||
| AF065314.1 | AAC17440.1 | |||
| AK131300.1 | BAD18468.1 | |||
| AK308149.1 | ||||
| BC096298.1 | AAH96298.1 | |||
| BC096299.1 | AAH96299.1 | |||
| BC096300.1 | AAH96300.1 | |||
| DR002103.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv518056 | Chr.2:97970379 - 98539833 on Build GRCh38 | Gain |
|
| nsv1013617 | Chr.2:98326914 - 98387710 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map