Assay Details
Target Gene Details
Entrez Gene ID: | 29925 |
Gene Name: | GDP-mannose pyrophosphorylase B |
Gene Aliases: |
MDDGA14, MDDGB14, MDDGC14 |
Location: |
Chr.3:49721476-49723974 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GMPPB | NM_013334.3 | NP_037466.2 | ||
| NM_021971.2 | NP_068806.1 | |||
| AK024319.1 | 8 | 1728 | BAB14882.1 | |
| AK026582.1 | 1 | 449 | ||
| AK291700.1 |
Target Gene Details
Entrez Gene ID: | 63891 |
Gene Name: | ring finger protein 123 |
Gene Aliases: |
FP1477, KPC1 |
Location: |
Chr.3:49688753-49721529 on Build GRCh38 |
Assay Gene Location: | Within Exon 41 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF123 | NM_022064.4 | 39 | 4197 | NP_071347.2 |
| NR_135218.1 | 39 | 4437 | ||
| XM_006713285.1 | 39 | 4355 | XP_006713348.1 | |
| XM_011533995.1 | 39 | 4132 | XP_011532297.1 | |
| XM_017007018.1 | 39 | 4159 | XP_016862507.1 | |
| AF370367.1 | 30 | 3351 | AAQ15203.1 | |
| AK022627.1 | 17 | 2023 | BAB14139.1 | |
| AK026968.1 | BAB15607.1 | |||
| AK090632.1 | 29 | 3239 | ||
| AK096353.1 | 39 | 4111 | ||
| AK126883.1 | 23 | 3904 | ||
| AL136729.1 | 12 | 1581 | CAB66663.2 | |
| AL832034.1 | 39 | 4159 | ||
| AL832138.1 | 38 | 4448 | ||
| AL832344.1 | 39 | 4135 | ||
| AY744152.1 | 38 | 4080 | AAU93470.1 | |
| BC041145.1 | 24 | 2730 | AAH41145.1 | |
| BC057392.1 | 26 | 2887 | AAH57392.2 | |
| BC088801.1 | 21 | 2458 | AAH88801.1 | |
| CR627051.1 | 35 | 5769 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508217 | Chr.3:49578013 - 49732463 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map