Assay Details
Target Gene Details
Entrez Gene ID: | 57109 |
Gene Name: | REX4 homolog, 3'-5' exonuclease |
Gene Aliases: |
REX4, XPMC2, XPMC2H |
Location: |
Chr.9:133406058-133418172 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| REXO4 | NM_001279349.1 | 6 | 1005 | NP_001266278.1 |
| NM_001279350.1 | 8 | 1343 | NP_001266279.1 | |
| NM_001279351.1 | 8 | 1204 | NP_001266280.1 | |
| NM_020385.3 | 8 | 1521 | NP_065118.2 | |
| NR_103995.1 | 6 | 980 | ||
| NR_103996.1 | 7 | 1174 | ||
| AF273304.1 | 8 | 1469 | AAF98162.1 | |
| AF295774.1 | 8 | 1469 | AAG02123.1 | |
| AK000247.1 | 5 | 382 | ||
| AK025493.1 | 8 | 1375 | BAB15152.1 | |
| AK295982.1 | 8 | 1204 | ||
| AK304547.1 | 8 | 1259 | ||
| AK310164.1 | ||||
| AK314333.1 | ||||
| AK315904.1 | 9 | 1194 | ||
| AL136894.1 | 8 | 1452 | CAB66828.1 | |
| BC009274.2 | 8 | 1424 | AAH09274.1 | |
| BX333016.2 | 6 | 894 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion |
|
| nsv433408 | Chr.9:133399364 - 133412362 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map