Assay Details
Target Gene Details
Entrez Gene ID: | 267 |
Gene Name: | autocrine motility factor receptor |
Gene Aliases: |
GP78, RNF45 |
Location: |
Chr.16:56361452-56425538 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMFR | NM_001144.5 | 14 | 2774 | NP_001135.3 |
| NM_001323511.1 | 14 | 2393 | NP_001310440.1 | |
| NM_001323512.1 | 15 | 2870 | NP_001310441.1 | |
| XM_005255890.3 | 14 | 2834 | XP_005255947.1 | |
| AF124145.1 | 14 | 2628 | AAD56722.1 | |
| AK023874.1 | 7 | 1531 | ||
| AK123962.1 | 1 | 1568 | ||
| BC017043.1 | 7 | 1511 | AAH17043.1 | |
| BC056869.1 | 5 | 1240 | AAH56869.1 | |
| BC069197.1 | 14 | 2774 | AAH69197.1 | |
| BX345849.2 | 1 | 462 | ||
| CA425340.1 | 1 | 317 | ||
| L35233.1 | 7 | 1506 | AAA79362.1 | |
| M63175.1 | 3 | 1014 | AAA36671.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833242 | Chr.16:56259354 - 56446840 on Build GRCh38 | Loss |
|
| nsv525929 | Chr.16:56350808 - 56367064 on Build GRCh38 | Loss |
|
| nsv519979 | Chr.16:56306184 - 56367064 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map