Assay Details
Target Gene Details
Entrez Gene ID: | 6311 |
Gene Name: | ataxin 2 |
Gene Aliases: |
ASL13, ATX2, SCA2, TNRC13 |
Location: |
Chr.12:111452214-111599676 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATXN2 | NM_001310121.1 | 17 | 2219 | NP_001297050.1 |
| NM_001310123.1 | 15 | 2110 | NP_001297052.1 | |
| NM_002973.3 | 17 | 3013 | NP_002964.3 | |
| NR_132311.1 | 17 | 3013 | ||
| BC111757.1 | 17 | 2219 | AAI11758.1 | |
| BC114546.1 | 15 | 2110 | AAI14547.1 | |
| U70323.1 | 17 | 3010 | AAB19200.1 | |
| Y08262.1 | 17 | 2610 | CAA69589.1 |
More Information
Set Membership: |
|

Genomic Map