Assay Details
Target Gene Details
Entrez Gene ID: | 22824 |
Gene Name: | heat shock protein family A (Hsp70) member 4 like |
Gene Aliases: |
APG-1, HSPH3, Osp94 |
Location: |
Chr.4:127781821-127834073 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPA4L | NM_001317381.1 | NP_001304310.1 | ||
| NM_001317383.1 | 1 | 279 | NP_001304312.1 | |
| NM_014278.3 | 1 | 279 | NP_055093.2 | |
| XM_011531745.2 | 1 | 348 | XP_011530047.1 | |
| AB023421.1 | BAA75063.1 | |||
| AK027822.1 | 1 | 166 | ||
| AK303053.1 | ||||
| AK307464.1 | 1 | 166 | ||
| AY290862.1 | 1 | 145 | AAP44471.1 | |
| BC040560.1 | 1 | 190 | AAH40560.1 | |
| DB051636.1 | 1 | 279 | ||
| EF197155.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv199n111 | Chr.4:127778281 - 127796436 on Build GRCh38 | Duplication |
|
| nsv1161147 | Chr.4:127775589 - 127818071 on Build GRCh38 | Duplication |
|
| nsv437935 | Chr.4:127768781 - 127782795 on Build GRCh38 | Loss |
|
| nsv830057 | Chr.4:127670133 - 127867622 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map