Assay Details
Target Gene Details
Entrez Gene ID: | 4189 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member B9 |
Gene Aliases: |
ERdj4, MDG-1, MDG1, MST049, MSTP049 |
Location: |
Chr.7:108569745-108574850 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJB9 | NM_012328.2 | NP_036460.1 | ||
| AB026908.1 | BAA84703.1 | |||
| AF083247.1 | AAD39845.1 | |||
| AF093824.1 | AAP97220.1 | |||
| AK092204.1 | ||||
| AL080081.1 | CAB45701.1 | |||
| BC028912.1 | AAH28912.1 | |||
| DA202575.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3572052 | Chr.7:108554229 - 108651992 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs150342729] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map