Assay Details
Target Gene Details
Entrez Gene ID: | 79962 |
Gene Name: | DnaJ heat shock protein family (Hsp40) member C22 |
Gene Aliases: |
wus |
Location: |
Chr.12:49346917-49353684 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNAJC22 | NM_001304944.1 | 3 | 1344 | NP_001291873.1 |
| NM_024902.3 | 2 | 882 | NP_079178.2 | |
| XM_005269155.2 | 2 | 393 | XP_005269212.1 | |
| XM_005269156.2 | 2 | 393 | XP_005269213.1 | |
| XM_005269157.2 | 2 | 393 | XP_005269214.1 | |
| AK055747.1 | 3 | 1344 | ||
| BC033236.1 | 2 | 541 | AAH33236.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832405 | Chr.12:49326496 - 49517079 on Build GRCh38 | Gain |
|
| dgv1497n100 | Chr.12:49284746 - 49438835 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map