Assay Details
Target Gene Details
Entrez Gene ID: | 286133 |
Gene Name: | scavenger receptor class A member 5 |
Gene Aliases: |
NET33, Tesr |
Location: |
Chr.8:27869882-27992852 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCARA5 | NM_173833.5 | 9 | 3666 | NP_776194.2 |
| AK172746.1 | 9 | 3496 | BAD18733.1 | |
| AY358150.1 | 8 | 2814 | AAQ88517.1 |
More Information
Set Membership: |
|

Genomic Map