Assay Details
Target Gene Details
Entrez Gene ID: | 84938 |
Gene Name: | autophagy related 4C cysteine peptidase |
Gene Aliases: |
APG4-C, APG4C, AUTL1, AUTL3 |
Location: |
Chr.1:62784106-62865270 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATG4C | NM_032852.3 | 1 | 77 | NP_116241.2 |
| NM_178221.2 | NP_835739.1 | |||
| XM_011542313.2 | 1 | 51 | XP_011540615.1 | |
| XM_017002600.1 | 1 | 51 | XP_016858089.1 | |
| XM_017002601.1 | XP_016858090.1 | |||
| AK027773.1 | 1 | 48 | BAB55356.1 | |
| BC018678.1 | 1 | 35 | ||
| BC033024.1 | 1 | 36 | AAH33024.1 | |
| BI518482.1 | 1 | 62 | ||
| DB183325.1 |
More Information
Set Membership: |
|

Genomic Map