Assay Details
Target Gene Details
Entrez Gene ID: | 3787 |
Gene Name: | potassium voltage-gated channel modifier subfamily S member 1 |
Gene Aliases: |
Kv9.1 |
Location: |
Chr.20:45091214-45101112 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNS1 | NM_001322799.1 | 3 | 841 | NP_001309728.1 |
| NM_002251.4 | 4 | 1043 | NP_002242.2 | |
| XM_017027846.1 | 1 | 1582 | XP_016883335.1 | |
| AF043473.2 | 4 | 1086 | AAC13165.2 | |
| AK289735.1 | 3 | 838 | ||
| BC075033.2 | 3 | 673 | AAH75033.2 | |
| BC075034.2 | 3 | 673 | AAH75034.2 | |
| BC132957.1 | 3 | 696 | ||
| BC132959.1 | 3 | 696 | ||
| BC144234.1 | 3 | 696 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map