Assay Details
Target Gene Details
Entrez Gene ID: | 10887 |
Gene Name: | prokineticin receptor 1 |
Gene Aliases: |
GPR73, GPR73a, PKR1, ZAQ |
Location: |
Chr.2:68645822-68655576 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PROKR1 | NM_138964.2 | 1 | 158 | NP_620414.1 |
| AB084080.1 | 1 | 158 | BAC24021.1 | |
| AF506287.1 | 1 | 158 | AAM48127.1 | |
| AY089976.1 | 1 | 158 | AAM11892.1 | |
| BC100960.2 | 1 | 158 | AAI00961.1 | |
| EF577399.1 | 1 | 158 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003458 | Chr.2:68617209 - 68676278 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map