Assay Details
Target Gene Details
Entrez Gene ID: | 84868 |
Gene Name: | hepatitis A virus cellular receptor 2 |
Gene Aliases: |
CD366, HAVcr-2, KIM-3, TIM3, TIMD-3, TIMD3, Tim-3 |
Location: |
Chr.5:157085832-157109237 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HAVCR2 | NM_032782.4 | 2 | 457 | NP_116171.3 |
| AB924452.1 | 1 | 140 | ||
| AF251707.1 | 2 | 329 | AAM19100.1 | |
| AF450242.1 | 2 | 260 | AAL65157.1 | |
| AF450243.1 | 2 | 260 | AAL65158.1 | |
| AK027334.1 | 2 | 246 | BAB55044.1 | |
| AK314406.1 | 2 | 264 | ||
| AY069944.1 | 2 | 203 | AAL55401.1 | |
| BC020843.1 | 2 | 312 | AAH20843.1 | |
| BC063431.1 | 2 | 347 | AAH63431.1 | |
| BP291914.1 | 2 | 457 | ||
| JX049979.1 | 2 | 203 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1109742 | Chr.5:157099986 - 157110561 on Build GRCh38 | Deletion |
|
| esv3575892 | Chr.5:156975204 - 157220778 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map