Assay Details
Target Gene Details
Entrez Gene ID: | 171177 |
Gene Name: | ras homolog family member V |
Gene Aliases: |
ARHV, CHP, WRCH2 |
Location: |
Chr.15:40872214-40874289 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RHOV | NM_133639.3 | 3 | 599 | NP_598378.3 |
| AB079131.1 | 3 | 551 | BAB86363.1 | |
| AY059636.1 | 3 | 449 | AAL17966.1 | |
| BC019353.1 | 3 | 529 | ||
| BC105020.1 | 3 | 539 | AAI05021.1 | |
| BC105022.1 | 3 | 539 | AAI05023.1 | |
| BC112945.1 | 3 | 525 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054764 | Chr.15:40776427 - 40904212 on Build GRCh38 | Gain |
|
| nsv569228 | Chr.15:40814287 - 40901953 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map