Assay Details
Target Gene Details
Entrez Gene ID: | 835 |
Gene Name: | caspase 2 |
Gene Aliases: |
CASP-2, ICH1, NEDD-2, NEDD2, PPP1R57 |
Location: |
Chr.7:143288215-143307696 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASP2 | NM_001224.4 | 12 | 3985 | NP_001215.1 |
| NM_032982.3 | 11 | 4170 | NP_116764.2 | |
| NM_032983.3 | 10 | 3952 | NP_116765.2 | |
| AK096245.1 | 1 | 1626 | ||
| BQ447606.1 | 1 | 73 | ||
| BX537669.1 | 1 | 5669 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831169 | Chr.7:143188183 - 143348025 on Build GRCh38 | Loss |
|
| nsv5984 | Chr.7:143305004 - 143350170 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map