Assay Details
Target Gene Details
Entrez Gene ID: | 4985 |
Gene Name: | opioid receptor delta 1 |
Gene Aliases: |
OPRD |
Location: |
Chr.1:28812142-28863696 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OPRD1 | NM_000911.3 | 2 | 555 | NP_000902.3 |
| EF057065.1 | 2 | 313 | ||
| EU883570.1 | 2 | 313 | ||
| U07882.2 | 2 | 546 | AAA18789.2 | |
| U10504.1 | 2 | 313 | AAA83426.1 |
More Information
Set Membership: |
|

Genomic Map