Assay Details
Target Gene Details
Entrez Gene ID: | 1837 |
Gene Name: | dystrobrevin alpha |
Gene Aliases: |
D18S892E, DRP3, DTN, DTN-A, LVNC1 |
Location: |
Chr.18:34493290-34891844 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DTNA | NM_001198945.1 | 1 | 152 | NP_001185874.1 |
| NM_001392.4 | 1 | 152 | NP_001383.2 | |
| NM_032975.3 | 1 | 152 | NP_116757.2 | |
| NM_032979.4 | 1 | 152 | NP_116761.2 | |
| XM_017025576.1 | 1 | 141 | XP_016881065.1 | |
| XM_017025578.1 | 1 | 141 | XP_016881067.1 | |
| XM_017025581.1 | 1 | 141 | XP_016881070.1 | |
| XM_017025584.1 | 1 | 141 | XP_016881073.1 | |
| XM_017025585.1 | 1 | 141 | XP_016881074.1 | |
| XM_017025586.1 | 1 | 141 | XP_016881075.1 | |
| XM_017025587.1 | 1 | 141 | XP_016881076.1 | |
| XM_017025590.1 | 1 | 141 | XP_016881079.1 | |
| XM_017025593.1 | 1 | 141 | XP_016881082.1 | |
| XM_017025597.1 | 1 | 141 | XP_016881086.1 | |
| XM_017025601.1 | 1 | 141 | XP_016881090.1 | |
| AJ009668.1 | 1 | 62 | CAA08769.1 | |
| AK294746.1 | 1 | 138 | ||
| DA073612.1 | 1 | 152 | ||
| U26742.1 | 1 | 125 | AAC50424.1 |
More Information
Set Membership: |
|

Genomic Map