Assay Details
Target Gene Details
Entrez Gene ID: | 1040 |
Gene Name: | CDP-diacylglycerol synthase 1 |
Gene Aliases: |
CDS |
Location: |
Chr.4:84582904-84651340 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDS1 | NM_001263.3 | 1 | 331 | NP_001254.2 |
| XM_005262687.2 | 1 | 327 | XP_005262744.1 | |
| XM_017007648.1 | 1 | 327 | XP_016863137.1 | |
| XM_017007649.1 | 1 | 327 | XP_016863138.1 | |
| XM_017007650.1 | 1 | 327 | XP_016863139.1 | |
| XM_017007651.1 | 1 | 327 | XP_016863140.1 | |
| AI673547.1 | ||||
| AK314245.1 | 1 | 68 | ||
| AU139278.1 | 1 | 337 | ||
| U60808.1 | AAC51184.1 | |||
| U65887.1 | 1 | 269 | AAC50735.1 |
More Information
Set Membership: |
|

Genomic Map