Assay Details
Target Gene Details
Entrez Gene ID: | 387129 |
Gene Name: | neuropeptide S receptor 1 |
Gene Aliases: |
ASRT2, GPR154, GPRA, NPSR, PGR14, VRR1 |
Location: |
Chr.7:34658239-34878332 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPSR1 | NM_001300933.1 | NP_001287862.1 | ||
| NM_001300934.1 | 7 | 1220 | NP_001287863.1 | |
| NM_001300935.1 | 10 | 1518 | NP_001287864.1 | |
| NM_207172.1 | 9 | 1372 | NP_997055.1 | |
| NM_207173.1 | NP_997056.1 | |||
| AY310326.1 | 9 | 1372 | AAQ76966.1 | |
| AY310327.1 | AAQ76967.1 | |||
| AY310328.1 | AAQ76968.1 | |||
| AY310330.1 | 8 | 1268 | AAQ76970.1 | |
| AY310331.1 | 8 | 1278 | AAQ76971.1 | |
| AY310332.1 | 7 | 1174 | AAQ76972.1 | |
| BC132693.1 | ||||
| BC132695.1 | ||||
| BC144380.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1080e201 | Chr.7:32621750 - 35008557 on Build GRCh38 | Deletion |
|
| nsv1020423 | Chr.7:34807572 - 34850088 on Build GRCh38 | Gain |
|
| dgv418n21 | Chr.7:34849610 - 34884725 on Build GRCh38 | Loss |
|
| nsv1135982 | Chr.7:32794601 - 34893593 on Build GRCh38 | Deletion |
|
| nsv436907 | Chr.7:32668474 - 34999437 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map