Assay Details
Target Gene Details
Entrez Gene ID: | 6286 |
Gene Name: | S100 calcium binding protein P |
Gene Aliases: |
MIG9 |
Location: |
Chr.4:6693839-6697170 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100P | NM_005980.2 | NP_005971.1 | ||
| AF539739.1 | AAO41114.1 | |||
| AI202505.1 | ||||
| BC006819.1 | AAH06819.1 | |||
| BI259919.1 | ||||
| X65614.1 | CAA46566.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829849 | Chr.4:6503821 - 6698228 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map