Assay Details
Target Gene Details
Entrez Gene ID: | 23314 |
Gene Name: | SATB homeobox 2 |
Gene Aliases: |
GLSS |
Location: |
Chr.2:199269500-199471266 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SATB2 | NM_015265.3 | NP_056080.1 | ||
| AK291463.1 |
Target Gene Details
Entrez Gene ID: | 150538 |
Gene Name: | SATB2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.2:199468098-199472758 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SATB2-AS1 | NR_026830.1 | 2 | 749 | |
| AK056625.1 | 1 | 296 | BAB71234.1 | |
| AK123639.1 | 2 | 762 | ||
| AK290476.1 | 1 | 196 | ||
| AK291368.1 | 1 | 95 |
More Information
Set Membership: |
|

Genomic Map