Assay Details
Target Gene Details
Entrez Gene ID: | 64761 |
Gene Name: | poly(ADP-ribose) polymerase family member 12 |
Gene Aliases: |
ARTD12, MST109, MSTP109, ZC3H1, ZC3HDC1 |
Location: |
Chr.7:140023744-140063721 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARP12 | NM_022750.3 | NP_073587.1 | ||
| NR_130117.1 | ||||
| XM_011516491.2 | XP_011514793.1 | |||
| AK026346.1 | BAB15457.1 | |||
| AL136766.1 | CAB66700.1 | |||
| AL137255.1 | CAB70657.1 | |||
| BC081541.1 | AAH81541.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2735212 | Chr.7:138060002 - 140415947 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs118130826] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map