Assay Details
Target Gene Details
Entrez Gene ID: | 7180 |
Gene Name: | cysteine rich secretory protein 2 |
Gene Aliases: |
CRISP-2, CT36, GAPDL5, TPX1, TSP1 |
Location: |
Chr.6:49676665-49714198 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3608949 | Chr.6:49690413 - 49880115 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map