Assay Details
Target Gene Details
Entrez Gene ID: | 89882 |
Gene Name: | tumor protein D52-like 3 |
Gene Aliases: |
NYDSP25, hD55 |
Location: |
Chr.9:6328349-6331900 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TPD52L3 | NM_001001874.2 | 2 | 1455 | NP_001001874.1 |
| XM_017015280.1 | 3 | 1511 | XP_016870769.1 | |
| BC033792.1 | 2 | 1400 | AAH33792.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1029665 | Chr.9:5687332 - 6775107 on Build GRCh38 | Gain |
|
| nsv613205 | Chr.9:6032924 - 6332901 on Build GRCh38 | Loss |
|
| dgv7387n100 | Chr.9:6308178 - 6408875 on Build GRCh38 | Gain |
|
| dgv7386n100 | Chr.9:6188652 - 6687205 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map