Assay Details
Target Gene Details
Entrez Gene ID: | 23179 |
Gene Name: | ral guanine nucleotide dissociation stimulator like 1 |
Gene Aliases: |
RGL |
Location: |
Chr.1:183636047-183928551 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGL1 | NM_001297669.1 | NP_001284598.1 | ||
| NM_001297670.1 | NP_001284599.1 | |||
| NM_015149.4 | NP_055964.3 | |||
| XM_011509339.2 | XP_011507641.1 | |||
| XM_011509341.1 | XP_011507643.1 | |||
| XM_011509342.2 | XP_011507644.1 | |||
| AF186780.1 | AAF67281.1 | |||
| AK295179.1 | ||||
| AK295611.1 | ||||
| AK316526.1 | ||||
| BC136591.1 | ||||
| BP235344.1 | ||||
| BX537416.1 | CAD97658.1 | |||
| DC333918.1 |
More Information
Set Membership: |
|

Genomic Map