Assay Details
Target Gene Details
Entrez Gene ID: | 218 |
Gene Name: | aldehyde dehydrogenase 3 family member A1 |
Gene Aliases: |
ALDH3, ALDHIII |
Location: |
Chr.17:19737984-19748433 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH3A1 | NM_000691.4 | NP_000682.3 | ||
| NM_001135167.1 | NP_001128639.1 | |||
| NM_001135168.1 | NP_001128640.1 | |||
| XM_005256522.1 | XP_005256579.1 | |||
| XM_005256523.2 | XP_005256580.1 | |||
| XM_005256524.3 | XP_005256581.1 | |||
| XM_011523730.1 | XP_011522032.1 | |||
| XM_011523731.1 | XP_011522033.1 | |||
| AK091272.1 | 1 | 955 | ||
| AK093755.1 | ||||
| AK093877.1 | BAC04239.1 | |||
| AK225505.1 | ||||
| AK225513.1 | ||||
| AK292193.1 | ||||
| AK314584.1 | ||||
| BC004102.2 | AAH04102.1 | |||
| BC004370.1 | AAH04370.1 | |||
| BC008892.2 | AAH08892.1 | |||
| BC021194.2 | AAH21194.1 | |||
| BM979497.1 | ||||
| BT007102.1 | AAP35766.1 | |||
| CV575863.1 | ||||
| DA427150.1 | ||||
| M74542.1 | AAA51696.1 | |||
| M77477.1 | AAB46377.1 | |||
| S61044.1 | AAB26658.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1063892 | Chr.17:19661321 - 19905172 on Build GRCh38 | Gain |
|
| nsv470582 | Chr.17:19717164 - 19760434 on Build GRCh38 | Loss |
|
| nsv953846 | Chr.17:19738588 - 19750287 on Build GRCh38 | Deletion |
|
| nsv526290 | Chr.17:19710044 - 19748169 on Build GRCh38 | Loss |
|
| nsv1061317 | Chr.17:19047542 - 20322438 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map