Assay Details
Target Gene Details
Entrez Gene ID: | 54925 |
Gene Name: | zinc finger and SCAN domain containing 32 |
Gene Aliases: |
HCCS-5, ZNF434 |
Location: |
Chr.16:3382081-3401065 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZSCAN32 | NM_001284527.1 | NP_001271456.1 | ||
| NM_001284528.1 | NP_001271457.1 | |||
| NM_001284529.1 | NP_001271458.1 | |||
| NM_001324340.1 | NP_001311269.1 | |||
| NM_001324341.1 | NP_001311270.1 | |||
| NM_001324342.1 | NP_001311271.1 | |||
| NM_001324343.1 | NP_001311272.1 | |||
| NM_001324344.1 | NP_001311273.1 | |||
| NM_001324345.1 | NP_001311274.1 | |||
| NM_001324346.1 | NP_001311275.1 | |||
| NM_017810.3 | NP_060280.2 | |||
| XM_005255403.2 | XP_005255460.1 | |||
| XM_011522555.2 | XP_011520857.1 | |||
| XM_017023371.1 | XP_016878860.1 | |||
| XM_017023372.1 | XP_016878861.1 | |||
| XM_017023373.1 | XP_016878862.1 | |||
| AK056463.1 | ||||
| AK291213.1 | ||||
| AK299070.1 | 4 | 870 | ||
| AK301586.1 | ||||
| AK303338.1 | 4 | 720 | ||
| AY261676.1 | AAP57398.1 | |||
| AY279347.1 | AAQ62844.1 | |||
| AY279348.1 | 5 | 1400 | AAQ62845.1 | |
| BC002859.2 | AAH02859.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1044147 | Chr.16:3149392 - 3392037 on Build GRCh38 | Loss |
|
| nsv1054969 | Chr.16:3358911 - 3391256 on Build GRCh38 | Loss |
|
| dgv2695n100 | Chr.16:3347901 - 3424504 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map