Assay Details
Target Gene Details
Entrez Gene ID: | 6720 |
Gene Name: | sterol regulatory element binding transcription factor 1 |
Gene Aliases: |
SREBP-1c, SREBP1, SREBP1a, bHLHd1 |
Location: |
Chr.17:17811349-17837017 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SREBF1 | NM_001005291.2 | 20 | 3934 | NP_001005291.1 |
| NM_001321096.2 | 19 | 3706 | NP_001308025.1 | |
| NM_004176.4 | 19 | 3844 | NP_004167.3 | |
| XM_005256772.4 | 19 | 3844 | XP_005256829.1 | |
| XM_017024970.1 | 17 | 3275 | XP_016880459.1 | |
| XM_017024971.1 | 15 | 2626 | XP_016880460.1 | |
| AB209609.1 | 18 | 4310 | BAD92846.1 | |
| AK091131.1 | 19 | 3673 | ||
| AK095325.1 | 14 | 2683 | ||
| AK128320.1 | 17 | 3353 | ||
| AK293795.1 | 19 | 3544 | ||
| AK294800.1 | 20 | 3522 | ||
| AK297113.1 | 17 | 3258 | ||
| BC023621.2 | 13 | 2268 | AAH23621.1 | |
| BC026962.1 | 13 | 2460 | AAH26962.1 | |
| BC057388.1 | 19 | 3835 | AAH57388.1 | |
| BC063281.1 | 20 | 3909 | AAH63281.1 | |
| BE208013.1 | 1 | 351 | ||
| U00968.1 | 19 | 3816 | AAC50051.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2659754 | Chr.17:17806631 - 17812878 on Build GRCh38 | Deletion |
|
| nsv953838 | Chr.17:17700587 - 17819486 on Build GRCh38 | Deletion |
|
| nsv522668 | Chr.17:17716545 - 17849495 on Build GRCh38 | Loss |
|
| nsv833385 | Chr.17:17690723 - 17862264 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map