Assay Details
Target Gene Details
Entrez Gene ID: | 440822 |
Gene Name: | piwi like RNA-mediated gene silencing 3 |
Gene Aliases: |
HIWI3 |
Location: |
Chr.22:24719034-24774720 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIWIL3 | NM_001008496.3 | NP_001008496.2 | ||
| NM_001255975.1 | NP_001242904.1 | |||
| NR_045648.1 | ||||
| NR_045649.1 | ||||
| AB079368.1 | BAC81343.1 | |||
| AK302409.1 | ||||
| AK315806.1 |
More Information
Set Membership: |
|

Genomic Map