Assay Details
Target Gene Details
Entrez Gene ID: | 3229 |
Gene Name: | homeobox C13 |
Gene Aliases: |
ECTD9, HOX3, HOX3G |
Location: |
Chr.12:53938792-53946544 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOXC13 | NM_017410.2 | NP_059106.2 | ||
| AF255676.1 | AAF67760.1 | |||
| AF263466.2 | AAF73439.1 | |||
| AK024027.1 | BAB14786.1 | |||
| BC090850.1 | AAH90850.1 | |||
| BT009908.1 | AAP88910.1 | |||
| BU158600.1 |
Target Gene Details
Entrez Gene ID: | 100874366 |
Gene Name: | HOXC13 antisense RNA |
Gene Aliases: |
HOXC-AS5 |
Location: |
Chr.12:53935328-53939643 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HOXC13-AS | NR_047507.1 | |||
| CN364530.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv826379 | Chr.12:53935085 - 54056069 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map