Assay Details
Target Gene Details
Entrez Gene ID: | 51005 |
Gene Name: | amidohydrolase domain containing 2 |
Gene Aliases: |
CGI-14 |
Location: |
Chr.16:2520351-2531414 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AMDHD2 | NM_001145815.1 | NP_001139287.1 | ||
| NM_015944.3 | 10 | 1510 | NP_057028.2 | |
| XM_017023262.1 | 11 | 1431 | XP_016878751.1 | |
| XM_017023263.1 | 9 | 1419 | XP_016878752.1 | |
| XM_017023264.1 | 10 | 1362 | XP_016878753.1 | |
| XM_017023265.1 | 10 | 1329 | XP_016878754.1 | |
| XM_017023266.1 | 10 | 1377 | XP_016878755.1 | |
| XM_017023267.1 | 9 | 1373 | XP_016878756.1 | |
| AB209351.1 | BAD92588.1 | |||
| AF132948.1 | AAD27723.1 | |||
| AK296877.1 | ||||
| AK300747.1 | ||||
| BC018734.1 | 10 | 1446 | AAH18734.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication |
|
| esv2760067 | Chr.16:2486824 - 2820535 on Build GRCh38 | Gain+Loss |
|
| nsv833122 | Chr.16:2501717 - 2663937 on Build GRCh38 | Loss |
|
| nsv1052260 | Chr.16:2074546 - 2629655 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75096863] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map