Assay Details
Target Gene Details
Entrez Gene ID: | 165904 |
Gene Name: | xin actin binding repeat containing 1 |
Gene Aliases: |
CMYA1, Xin |
Location: |
Chr.3:39183214-39192621 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| XIRP1 | NM_001198621.2 | 2 | 2698 | NP_001185550.1 |
| NM_194293.2 | 2 | 2688 | NP_919269.2 | |
| XM_005264909.3 | 2 | 2647 | XP_005264966.1 | |
| AJ626899.1 | 2 | 2696 | CAF25191.1 | |
| AJ626900.1 | 2 | 2696 | CAF25192.1 | |
| AK096421.1 | BAC04783.1 | |||
| AK122723.1 | 1 | 95 | ||
| AK126299.1 | 1 | 1138 | BAC86519.1 | |
| AL713648.1 | 1 | 776 | CAD28459.1 | |
| AL831956.1 | 1 | 483 | ||
| AL832020.1 | 1 | 776 | ||
| AL832459.1 | 1 | 776 | ||
| AY375160.1 | 2 | 2688 | AAQ64003.1 | |
| BC127119.1 | ||||
| BC139782.1 | ||||
| BX648565.1 | ||||
| CR749430.1 | 2 | 2698 | CAH18268.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv8287n54 | Chr.3:39146691 - 39189506 on Build GRCh38 | Loss |
|
| nsv590106 | Chr.3:39149538 - 39233827 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map