Assay Details
Target Gene Details
Entrez Gene ID: | 90326 |
Gene Name: | THAP domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.1:6624645-6635586 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THAP3 | NM_138350.3 | 5 | 578 | NP_612359.2 |
| XM_005263532.3 | 6 | 2088 | XP_005263589.1 | |
| XM_011542400.2 | 5 | 1662 | XP_011540702.1 | |
| XM_017002761.1 | 5 | 515 | XP_016858250.1 | |
| AK096217.1 | 5 | 536 | BAC04727.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs80089628] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map