Assay Details
Target Gene Details
Entrez Gene ID: | 11280 |
Gene Name: | sodium voltage-gated channel alpha subunit 11 |
Gene Aliases: |
FEPS3, HSAN7, NAV1.9, NaN, PN5, SCN12A, SNS-2 |
Location: |
Chr.3:38845764-39051986 on Build GRCh38 |
Assay Gene Location: | Within Exon 36 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCN11A | NM_001287223.1 | 27 | 4425 | NP_001274152.1 |
| NM_014139.2 | 26 | 4594 | NP_054858.2 | |
| XM_011533321.2 | 23 | 3819 | XP_011531623.1 | |
| XM_017005647.1 | 31 | 5046 | XP_016861136.1 | |
| XM_017005648.1 | 28 | 4527 | XP_016861137.1 | |
| XM_017005649.1 | 30 | 4924 | XP_016861138.1 | |
| XM_017005650.1 | 27 | 4725 | XP_016861139.1 | |
| XM_017005651.1 | 27 | 4251 | XP_016861140.1 | |
| XM_017005653.1 | 17 | 3203 | XP_016861142.1 | |
| AF109737.1 | 26 | 4594 | AAF24976.1 | |
| AF188679.1 | 27 | 4425 | AAF17480.1 | |
| AJ417790.1 | 26 | 4395 | CAD10507.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3595916 | Chr.3:38839386 - 38852617 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map