Assay Details
Target Gene Details
Entrez Gene ID: | 2178 |
Gene Name: | Fanconi anemia complementation group E |
Gene Aliases: |
FACE, FAE |
Location: |
Chr.6:35452339-35467106 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FANCE | NM_021922.2 | 2 | 971 | NP_068741.1 |
| XM_005248885.2 | 2 | 993 | XP_005248942.1 | |
| XM_005248886.2 | 2 | 993 | XP_005248943.1 | |
| XM_005248887.2 | 2 | 993 | XP_005248944.1 | |
| XM_005248888.3 | 2 | 993 | XP_005248945.1 | |
| XM_011514343.2 | 2 | 686 | XP_011512645.1 | |
| AF265210.1 | 2 | 971 | AAG16743.1 | |
| AK292522.1 | 2 | 982 | ||
| AK312617.1 | 2 | 959 | ||
| BC046359.1 | 2 | 977 | AAH46359.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830641 | Chr.6:35314757 - 35472826 on Build GRCh38 | Loss |
|
| nsv528351 | Chr.6:35410145 - 35498009 on Build GRCh38 | Loss |
|
| nsv602910 | Chr.6:35411001 - 35489619 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map