Assay Details
Target Gene Details
Entrez Gene ID: | 345274 |
Gene Name: | solute carrier family 10 member 6 |
Gene Aliases: |
SOAT |
Location: |
Chr.4:86823468-86849263 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC10A6 | NM_197965.2 | 1 | 300 | NP_932069.1 |
| AJ583502.1 | 1 | 152 | CAE47477.1 | |
| BC107051.1 | 1 | 152 | AAI07052.1 | |
| BC107052.1 | 1 | 152 | AAI07053.1 | |
| EF437223.1 | 1 | 300 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv594787 | Chr.4:86300674 - 86967515 on Build GRCh38 | Gain |
|
| nsv1013308 | Chr.4:86848541 - 86949016 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map