Assay Details
Target Gene Details
Entrez Gene ID: | 8540 |
Gene Name: | alkylglycerone phosphate synthase |
Gene Aliases: |
ADAP-S, ADAS, ADHAPS, ADPS, ALDHPSY, RCDP3 |
Location: |
Chr.2:177392743-177543836 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGPS | NM_003659.3 | NP_003650.1 | ||
| XM_011512041.2 | XP_011510343.1 | |||
| AK296239.1 | ||||
| AK309714.1 | ||||
| AK314259.1 | ||||
| AK316240.1 | ||||
| AY544121.1 | AAT11152.1 | |||
| BC141820.1 | ||||
| DC402978.1 | ||||
| Y09443.1 | CAA70591.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953527 | Chr.2:177393073 - 177407672 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map