Assay Details
Target Gene Details
Entrez Gene ID: | 85865 |
Gene Name: | GTP binding protein 10 |
Gene Aliases: |
ObgH2, UG0751c10 |
Location: |
Chr.7:90346665-90391455 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTPBP10 | NM_001042717.2 | NP_001036182.1 | ||
| NM_033107.3 | NP_149098.2 | |||
| AF351613.1 | AAN76513.1 | |||
| AK095561.1 | BAC04573.1 | |||
| AK294325.1 | ||||
| AL525056.3 | ||||
| BC004923.2 | AAH04923.3 | |||
| BC021573.1 | AAH21573.2 | |||
| BC107714.1 | AAI07715.1 | |||
| CR933597.1 | CAI45922.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891163 | Chr.7:90335679 - 90681223 on Build GRCh38 | Loss |
|
| esv24241 | Chr.7:90183329 - 90563872 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map