Assay Details
Target Gene Details
Entrez Gene ID: | 915 |
Gene Name: | CD3d molecule |
Gene Aliases: |
CD3-DELTA, IMD19, T3D |
Location: |
Chr.11:118339074-118344233 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD3D | NM_000732.4 | NP_000723.1 | ||
| NM_001040651.1 | NP_001035741.1 | |||
| XM_017018543.1 | XP_016874032.1 | |||
| AK311971.1 | ||||
| BC039035.1 | AAH39035.1 | |||
| BC070321.1 | AAH70321.1 | |||
| CD014058.1 | ||||
| JN392069.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3341931 | Chr.11:118339027 - 118340725 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map