Assay Details
Target Gene Details
Entrez Gene ID: | 6773 |
Gene Name: | signal transducer and activator of transcription 2 |
Gene Aliases: |
IMD44, ISGF-3, P113, STAT113 |
Location: |
Chr.12:56341597-56360253 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAT2 | NM_005419.3 | 24 | 4442 | NP_005410.1 |
| NM_198332.1 | 24 | 4430 | NP_938146.1 | |
| XM_011538697.2 | 24 | 4374 | XP_011536999.1 | |
| XM_011538698.2 | 24 | 4335 | XP_011537000.1 | |
| XM_011538700.2 | 18 | 3772 | XP_011537002.1 | |
| XM_017019904.1 | 23 | 4809 | XP_016875393.1 | |
| AK094039.1 | 1 | 2324 | ||
| BC051284.1 | 23 | 4262 | AAH51284.1 | |
| BX640607.1 | 11 | 3169 | CAE45713.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map