Assay Details
Target Gene Details
Entrez Gene ID: | 22978 |
Gene Name: | 5'-nucleotidase, cytosolic II |
Gene Aliases: |
GMP, NT5B, PNT5, SPG45, SPG65, cN-II |
Location: |
Chr.10:103088017-103193306 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7528 | Chr.10:103062273 - 103106919 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map