Assay Details
Target Gene Details
Entrez Gene ID: | 94059 |
Gene Name: | leukocyte receptor cluster member 9 |
Gene Aliases: |
- |
Location: |
Chr.19:54461796-54463711 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LENG9 | NM_001301782.1 | 1 | 472 | NP_001288711.1 |
| NM_198988.1 | 1 | 472 | NP_945339.2 | |
| BC015921.1 | 1 | 472 | AAH15921.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3663n100 | Chr.19:54289334 - 54592227 on Build GRCh38 | Gain |
|
| nsv953617 | Chr.19:54401196 - 54463717 on Build GRCh38 | Deletion |
|
| esv2758771 | Chr.19:54163847 - 54954956 on Build GRCh38 | Gain+Loss |
|
| nsv833878 | Chr.19:54268967 - 54518890 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map