Assay Details
Target Gene Details
Entrez Gene ID: | 11129 |
Gene Name: | CLK4 associating serine/arginine rich protein |
Gene Aliases: |
CLASP, SFRS16, SWAP2 |
Location: |
Chr.19:45039040-45070956 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLASRP | NM_001278439.1 | NP_001265368.1 | ||
| NM_007056.2 | NP_008987.2 | |||
| NR_103529.1 | ||||
| XM_011526396.2 | XP_011524698.1 | |||
| XM_011526397.2 | XP_011524699.1 | |||
| AF042800.1 | AAC82339.1 | |||
| AK094681.1 | ||||
| AK293333.1 | ||||
| AY358944.1 | AAQ89303.1 | |||
| BC080554.1 | AAH80554.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953587 | Chr.19:45039143 - 45166142 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map