Assay Details
Target Gene Details
Entrez Gene ID: | 56943 |
Gene Name: | ENY2, transcription and export complex 2 subunit |
Gene Aliases: |
DC6, Sus1, e(y)2 |
Location: |
Chr.8:109334323-109345960 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ENY2 | NM_001193557.1 | NP_001180486.1 | ||
| NM_020189.5 | NP_064574.1 | |||
| NR_036471.1 | ||||
| NR_036472.1 | ||||
| AF173296.1 | AAF89829.1 | |||
| AF201940.1 | AAF86876.1 | |||
| AK000487.1 | ||||
| AK095651.1 | ||||
| AK290752.1 | ||||
| AK316560.1 | ||||
| BC007870.2 | AAH07870.1 | |||
| BX648621.1 | ||||
| CR457183.1 | CAG33464.1 | |||
| DA548665.1 | ||||
| DA762166.1 | ||||
| DB168034.1 |
More Information
Set Membership: |
|

Genomic Map