Assay Details
Target Gene Details
Entrez Gene ID: | 27090 |
Gene Name: | ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 4 |
Gene Aliases: |
IV, SIAT3-C, SIAT3C, SIAT7-D, SIAT7D, ST6GALNACIV, ST6GalNAc |
Location: |
Chr.9:127907886-127917052 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST6GALNAC4 | NM_175039.3 | 6 | 1250 | NP_778204.1 |
| NM_175040.3 | 5 | 1163 | NP_778205.1 | |
| XM_011518527.2 | 7 | 1342 | XP_011516829.1 | |
| XM_011518529.2 | 7 | 1373 | XP_011516831.1 | |
| AB035172.1 | BAA87034.1 | |||
| AJ271734.1 | CAC07404.1 | |||
| AK000600.1 | 6 | 1246 | BAA91281.1 | |
| AK292049.1 | ||||
| BC036705.2 | 6 | 1156 | AAH36705.1 | |
| F27902.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv615381 | Chr.9:127906678 - 127954482 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map