Assay Details
Target Gene Details
Entrez Gene ID: | 23279 |
Gene Name: | nucleoporin 160 |
Gene Aliases: |
- |
Location: |
Chr.11:47778083-47848544 on Build GRCh38 |
Assay Gene Location: | Within Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUP160 | NM_015231.2 | 33 | 4060 | NP_056046.1 |
| NR_134636.1 | 33 | 4075 | ||
| AK304308.1 | 33 | 3846 | ||
| AL832029.1 | 33 | 3850 | ||
| AL832813.1 | 2 | 162 | ||
| BC008700.1 | 4 | 343 | AAH08700.1 | |
| BC125227.1 | 33 | 3983 | ||
| BC125228.1 | 33 | 3983 | ||
| BX647634.1 | 33 | 3860 | ||
| D83781.1 | 33 | 3829 | BAA12110.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469858 | Chr.11:47625963 - 47815620 on Build GRCh38 | Gain |
|
| dgv1111n100 | Chr.11:47653918 - 48644472 on Build GRCh38 | Gain |
|
| nsv832143 | Chr.11:47679256 - 47861988 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map