Assay Details
Target Gene Details
Entrez Gene ID: | 9844 |
Gene Name: | engulfment and cell motility 1 |
Gene Aliases: |
CED-12, CED12, ELMO-1 |
Location: |
Chr.7:36852906-37449409 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELMO1 | NM_001039459.2 | 7 | 1700 | NP_001034548.1 |
| NM_001206480.2 | 22 | 3479 | NP_001193409.1 | |
| NM_001206482.1 | 22 | 3649 | NP_001193411.1 | |
| NM_014800.10 | 22 | 3290 | NP_055615.8 | |
| NM_130442.3 | 7 | 1846 | NP_569709.1 | |
| NR_038120.1 | 6 | 1733 | ||
| XM_005249919.2 | 23 | 3843 | XP_005249976.1 | |
| XM_006715805.1 | 22 | 3135 | XP_006715868.1 | |
| XM_011515654.1 | 23 | 4528 | XP_011513956.1 | |
| XM_017012838.1 | 23 | 4713 | XP_016868327.1 | |
| XM_017012839.1 | 22 | 3077 | XP_016868328.1 | |
| AK126565.1 | 10 | 2330 | BAC86597.1 | |
| AK296253.1 | 20 | 2511 | ||
| AL136787.1 | 22 | 3191 | CAB66721.1 | |
| BC003051.1 | 7 | 1696 | AAH03051.1 | |
| BC064635.1 | 7 | 1648 | AAH64635.1 | |
| BC077074.1 | 7 | 1697 | AAH77074.1 | |
| D87457.1 | 7 | 1672 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5701 | Chr.7:36844883 - 36862375 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map