Assay Details
Target Gene Details
Entrez Gene ID: | 115265 |
Gene Name: | DNA damage inducible transcript 4 like |
Gene Aliases: |
REDD2, Rtp801L |
Location: |
Chr.4:100185870-100190498 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDIT4L | NM_145244.3 | NP_660287.1 | ||
| AK094236.1 | BAC04315.1 | |||
| AK312926.1 | ||||
| AL832277.1 | CAI46176.1 | |||
| BC013592.2 | AAH13592.1 | |||
| DB208869.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830019 | Chr.4:100102509 - 100287751 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map