Assay Details
Target Gene Details
Entrez Gene ID: | 387638 |
Gene Name: | chromosome 10 open reading frame 113 |
Gene Aliases: |
bA165O3.1 |
Location: |
Chr.10:21125763-21146559 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C10orf113 | NM_001010896.2 | 2 | 418 | NP_001010896.2 |
| NM_001177483.1 | 2 | 453 | NP_001170954.1 | |
| AI806552.1 | 1 | 166 | ||
| BC140744.1 | 2 | 418 |
Target Gene Details
Entrez Gene ID: | 10529 |
Gene Name: | nebulette |
Gene Aliases: |
LASP2, LNEBL |
Location: |
Chr.10:20779973-21174187 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEBL | NM_001173484.1 | NP_001166955.1 | ||
| NM_213569.2 | NP_998734.1 | |||
| AB209311.1 | BAD92548.1 | |||
| AJ580772.1 | CAE45323.1 | |||
| AK096540.1 | BAC04812.1 | |||
| AK295186.1 | ||||
| AK316521.1 | ||||
| BC126132.1 | ||||
| BC126134.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831806 | Chr.10:21084709 - 21230421 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map