Assay Details
Target Gene Details
Entrez Gene ID: | 3268 |
Gene Name: | ArfGAP with FG repeats 2 |
Gene Aliases: |
HRBL, RABR |
Location: |
Chr.7:100539211-100568220 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGFG2 | NM_006076.4 | NP_006067.3 | ||
| XM_005250306.2 | XP_005250363.1 | |||
| XM_017012112.1 | XP_016867601.1 | |||
| XM_017012113.1 | XP_016867602.1 | |||
| XM_017012114.1 | XP_016867603.1 | |||
| AF015042.1 | AAD01550.1 | |||
| AK314954.1 | ||||
| BC009393.2 | AAH09393.1 | |||
| BC017329.2 | AAH17329.1 | |||
| BF310674.1 | ||||
| BF314987.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map